Tetrabutylammonium tetrafluoroborate ≥98%

Supplier: Matrix Scientific

006518-5G
101779-146EA 15.38 USD
101779-146
Tetrabutylammonium tetrafluoroborate ≥98%
Tetrabutylammonium tetrafluoroborate

TCA listed: T
Hazard: Toxic/Hygroscopic

Formula: C₁₆H₃₆BF₄N
Melting Pt: 158…162 °C
MDL Number: MFCD00011634
CAS Number: 429-42-5

Order Now

Specification Test Results

Melting point 158-162°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR