4-tert-Butylbenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride

006402-25G 006402-100G
101779-050EA 32.31 USD
101779-050 101779-052
4-tert-Butylbenzenesulfonyl chloride ≥98%
4-tert-Butylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₁₀H₁₃ClO₂S
MW: 232.73
Boiling Pt: 165
Melting Pt: 83
MDL Number: MFCD00007449
CAS Number: 15084-51-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Melting point 77-78°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR