1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane

Supplier: Matrix Scientific

Synonyms: 1,3,5-Tris(3,3,3-trifluoropropyl)-1,3,5-trimethylcyclotrisiloxane, 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane

006184-25G 006184-5G
101778-932EA 67.69 USD
101778-932 101778-930
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane

TCA listed: T
Hazard: Toxic/Store in Freezer

Formula: C₁₂H₂₁F₉O₃Si₃
Boiling Pt: 135 °C (22 mmHg)
Melting Pt: 34…36 °C
Density: 1.24
MDL Number: MFCD00054667
CAS Number: 2374-14-3

Order Now

Specification Test Results

Boiling point 135°/22mm
Density 1.24


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR