3,5-Bis(trifluoromethyl)benzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene

005809-5G 005809-25G
101778-634EA 36.92 USD
101778-634 101778-636
3,5-Bis(trifluoromethyl)benzyl bromide ≥97%
3,5-Bis(trifluoromethyl)benzyl bromide

Hazard: Flammable/Corrosive/Refrigerate/Lachrymator

Formula: C₉H₅BrF₆
Boiling Pt: 45…47 °C (10 mmHg)
Density: 1.675
Flash Pt: 26 °C
MDL Number: MFCD00009905
CAS Number: 32247-96-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.675
ND20 1.445


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR