1,2-Difluoro-4-nitrobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 3,4-Difluoronitrobenzene

005547-500G
101778-474EA 169.23 USD
101778-474
1,2-Difluoro-4-nitrobenzene ≥98%
1,2-Difluoro-4-nitrobenzene

Hazard: Flammable

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80
Density: 1.437
MDL Number: MFCD00007198
CAS Number: 369-34-6
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 76-80°/11m
Density 1.437
ND20 1.509


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR