α,α,α-Trifluoro-m-toluoyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride

005455-25G 005455-100G
101778-338EA 27.69 USD
101778-338 101778-340
α,α,α-Trifluoro-m-toluoyl chloride ≥98%
α,α,α-Trifluoro-m-toluoyl chloride

TCA listed: T
Hazard: Corrosive/Lachrymator

Formula: C₈H₄ClF₃O
MW: 208.57
Boiling Pt: 112…114 °C (20 mmHg)
Density: 1.383
MDL Number: MFCD00000680
CAS Number: 2251-65-2
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Boiling point 112-114°/2
Density 1.383
ND20 1.477


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR