2-(Trifluoromethoxy)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethoxy)benzene-1-sulphonyl chloride, 2-(Trifluoromethoxy)benzene-1-sulfonyl chloride

005364-5G 005364-25G
101778-300EA 13.85 USD
101778-300 101778-302
2-(Trifluoromethoxy)benzenesulfonyl chloride ≥97%
2-(Trifluoromethoxy)benzenesulphonyl chloride

Hazard: Corrosive

Formula: C₇H₄ClF₃O₃S
Boiling Pt: 75…77 °C (1 mmHg)
Melting Pt: 29…32 °C
MDL Number: MFCD00052316
CAS Number: 103008-51-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 75-77°/1mm
ND20 1.4825


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR