2,6-Dichloroaniline ≥97%
Supplier: Matrix Scientific
005145-100G
005145-2.5KG
005145-500G
101777-978EA
21.54
USD
101777-978
101777-982
101777-980
2,6-Dichloroaniline ≥97%
2,6-Dichloroaniline
TCA listed: T
Hazard: Toxic/Irritant
Formula:
C₆H₅Cl₂N Melting Pt: 38…41 °C |
MDL Number:
MFCD00007675 CAS Number: 608-31-1 UN: 3442 ADR: 6.1,II |
Specification Test Results
Melting point | 38-41° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...