ω,4-Dibromoacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

004903-100G 004903-25G
101777-840EA 55.38 USD
101777-840 101777-838
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone

TCA listed: T
Hazard: Corrosive/Lachrymator

Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310
Melting Pt: 108…110 °C
MDL Number: MFCD00000200
CAS Number: 99-73-0
UN: 3261
ADR: 8 II
Merck Index: 13,01412

Order Now

Specification Test Results

Melting point 108-110°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR