Octafluoroadipic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
Perfluoroadipic Acid, Octafluorohexanedioic Acid, Perfluorohexanedioic Acid
004825-5G
004825-25G
004825-100G
101777-776EA
64.62
USD
101777-776
101777-778
101777-780
Octafluoroadipic acid ≥98%
Octafluoroadipic acid
Purity: 98%
CAS: 336-08-3
MDL Number: MFCD00066405
Molecular Formula: C6H2F8O4
Molecular Weight: 290,07
TCA listed: T
Hazard: Corrosive
Formula:
C₆H₂F₈O₄ Melting Pt: 132…134 °C |
MDL Number:
MFCD00066405 CAS Number: 336-08-3 |
Specification Test Results
Melting point | 132-134° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...