α,α,α-Trifluoro-3,5-diaminotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)-1,3-phenylenediamine, 5-(Trifluoromethyl)-m-phenylenediamine, 5-Trifluoromethyl-1,3-phenylenediamine, 5-(Trifluoromethyl)benzene-1,3-diamine, 3,5-Diaminobenzotrifluoride

004677-5G 004677-25G 004677-100G
101777-690EA 121.54 USD
101777-690 101777-692 101777-694
α,α,α-Trifluoro-3,5-diaminotoluene ≥98%
α,α,α-Trifluoro-3,5-diaminotoluene

Hazard: Toxic

Formula: C₇H₇F₃N₂
Melting Pt: 88…89 °C
MDL Number: MFCD00014776
CAS Number: 368-53-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 88-89°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR