2-(Tributylstannyl)pyridine ≥90%
Supplier: Matrix Scientific
Synonyms:
2-(Tri-n-butylstannyl)pyridine
004351-5G
101777-516EA
89.23
USD
101777-516
2-(Tributylstannyl)pyridine ≥90%
2-(Tributylstannyl)pyridine
Balance is tributyl halide
Hazard: Toxic/Irritant/Refrigerate
Formula:
C₁₇H₃₁NSn MW: 368.15 Boiling Pt: 130…132 °C (0.8 mmHg) Density: 1.14 |
MDL Number:
MFCD00052052 CAS Number: 17997-47-6 UN: 2788 ADR: 6.1,III |
Specification Test Results
Boiling point | 130-132°/0 |
Density | 1.14 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...