3-Bromothiophenol ≥98%

Supplier: Matrix Scientific

Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol

004210-25G
101777-442EA 273.85 USD
101777-442
3-Bromothiophenol ≥98%
3-Bromothiophenol

Hazard: Toxic/Corrosive/Stench

Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 123…124 °C (40 mmHg)
Density: 1.832
MDL Number: MFCD00009603
CAS Number: 6320-01-0
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 123-124°/4
Density 1.832
ND20 1.6324

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR