3-Chlorotetrafluoropropionyl chloride ≥97%

Supplier: Matrix Scientific

003496-1G 003496-5G
101776-764EA 1256.92 USD
101776-764 101776-766
3-Chlorotetrafluoropropionyl chloride ≥97%
3-Chlorotetrafluoropropionyl chloride

Hazard: Corrosive/Refrigerate

Formula: C₃Cl₂F₄O
MW: 198.93 g/mol
Boiling Pt: 49…51 °C
MDL Number: MFCD00153100
CAS Number: 24503-62-6

Order Now

Specification Test Results

Boiling point 49-51°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR