Tetrafluorotetracyanoquinodimethane ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile
003446-100MG
101776-728EA
283.08
USD
101776-728
Tetrafluorotetracyanoquinodimethane ≥95%
Tetrafluorotetracyanoquinodimethane
Purity: 95%
CAS: 29261-33-4
MDL Number: MFCD00042382
Molecular Formula: C12F4N4
Molecular Weight: 276,15
Hazard: Toxic
Formula:
C₁₂F₄N₄ MW: 276.15 g/mol Melting Pt: 291 °C |
MDL Number:
MFCD00042382 CAS Number: 29261-33-4 |
Specification Test Results
Melting point | 291° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...