3-Chlorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

003271-25G 003271-5G
101776-574EA 180 USD
101776-574 101776-572
3-Chlorobenzenesulfonyl chloride ≥98%
3-Chlorobenzenesulphonyl chloride

Hazard: Corrosive/Store in Freezer/Moisture Sensitive

Formula: C₆H₄Cl₂O₂S
Boiling Pt: 102…104 °C (1 mmHg)
Melting Pt: ∼ 21 °C
MDL Number: MFCD00051697
CAS Number: 2888-06-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 102-104°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR