3-[(Trifluoromethyl)thio]aniline ≥98%

Supplier: Matrix Scientific

003231-1G
101776-550EA 40 USD
101776-550
3-[(Trifluoromethyl)thio]aniline ≥98%
3-[(Trifluoromethyl)thio]aniline

Hazard: Toxic/Stench

Formula: C₇H₆F₃NS
Boiling Pt: 221 °C
Density: 1.342
MDL Number: MFCD00190130
CAS Number: 369-68-6

Order Now

Specification Test Results

Boiling point 221°
Density 1.342
ND20 1.522


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR