2-Chloro-6-fluoro-3-methylbenzoyl chloride

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-fluoro-3-methylbenzene-1-carbonyl chloride, 2-Chloro-6-fluoro-3-methylbenzoylchloride, 2-Chloro-6-fluoro-3-toluoyl chloride

002756-1G 002756-5G
101776-338EA 30.77 USD
101776-338 101776-340
2-Chloro-6-fluoro-3-methylbenzoyl chloride
2-Chloro-6-fluoro-3-methylbenzoyl chloride

Hazard: Corrosive

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
MDL Number: MFCD01631371
CAS Number: 261762-82-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR