1,4,7,10-Tetraazacyclododecane tetrahydrochloride

Supplier: Matrix Scientific

Synonyms: Cyclen tetrahydrochloride

001925-1G
101775-608EA 135.38 USD
101775-608
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

RTECS: XA5253000
Hazard: Irritant-Harmful/Moisture Sensitive

Formula: C₈H₂₄Cl₄N₄
MW: 318.12 g/mol
Melting Pt: 305 °C (dec.)
MDL Number: MFCD00068484
CAS Number: 10045-25-7

Order Now

Specification Test Results

Melting point 305°(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR