Perfluorobutane ≥97%

Supplier: Matrix Scientific

001836-100G
101775-558EA 1207.69 USD
101775-558
Perfluorobutane ≥97%
Perfluorobutane

TCA listed: T
Hazard: Nonflammable Gas/Store in Freezer

Formula: C₄F₁₀
Boiling Pt: –2 °C
Melting Pt: –84.5 °C
Density: 1.517
MDL Number: MFCD00042080
CAS Number: 355-25-9

Order Now

Specification Test Results

Boiling point -2°
Density 1.517


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR