1-Bromo-2-fluorobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Bromofluorobenzene

001728-100G 001728-500G
101775-522EA 84.62 USD
101775-522 101775-524
1-Bromo-2-fluorobenzene ≥99%
1-Bromo-2-fluorobenzene

TCA listed: T
Hazard: Flammable

Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155
Density: 1.601
MDL Number: MFCD00000282
CAS Number: 1072-85-1
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Boiling point 156°
Density 1.601
ND20 1.5339


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR