Pentafluorobenzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

001561-100G
101775-446EA 515.38 USD
101775-446
Pentafluorobenzyl bromide ≥98%
Pentafluorobenzyl bromide

Hazard: Corrosive/Irritant/Lachrymator

Formula: (C₆F₅)CH₂Br
Boiling Pt: 174…175 °C
Melting Pt: 18…20 °C
Density: 1.728
MDL Number: MFCD00000299
CAS Number: 1765-40-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 174-175°
Density 1.728
ND20 1.472


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR