Break-Safe™ Ampoule Opener, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60607
101765-012EA 107.59 USD
101765-012
Break-Safe™ Ampoule Opener, Electron Microscopy Sciences
Ampule Openers
EMS Introduces Break-Safe™ allowing you to safely open up to three ampoules at one time. Box style design allows one handed opening of up to 3 ampules at a time.
  • Transparency allows unobstructed viewing of ampule contents at all times
  • Copolyester plastic construction has broad chemical resistance
Gently pressing down on the lid snaps ampule necks clean while keeping them contained within the device and away from fingers. After removing ampule contents, simply turn the device upside down over a trash receptacle to safely discard ampules and tops.
Delivery information: Includes 4 inserts to allow opening of 1 mL, 2 mL, 5 mL and 10 mL ampules.