ProMax™ Syringe Filters, Dikma Technologies
Supplier: Dikma Technologies
37177 37180 37187 37178 37182 37185 37192 30009 30010 30011 30012 30013 30014 30015 30016 30017 30018 30019 30020
101764-758EA 137.31 USD
101764-758 101764-760 101764-764 101764-766 101764-768 101764-770 101764-772 101764-774 101764-776 101764-778 101764-780 101764-782 101764-784 101764-786 101764-788 101764-790 101764-792 101764-794 101764-796
ProMax™ Syringe Filters, Dikma Technologies
Filters Syringe Filters
Particulates can damage expensive equipment, valves, columns, and pumps. They can also lead to erratic analytical results. Pre-filtering samples prior to analysis is critical in preventing column and frit blockage, undue wear on valve seals, and abnormally high operating pressures.
  • Non sterile
  • Filters are available with CA, PVDF, PES, or PTFE membranes
The syringe filters are designed for economical and rapid filtration of almost any solution prior to analysis.
The syringe filters are available in a broad range of pore sizes and membrane types. Filters made with 13 or 25 mm membranes are suitable for use with samples 1 ml or greater.
The housing attaches to any standard Luer hub syringe, so the sample can be pushed through the membrane under pressure. The resulting eluent is free from particulates and ready for use with HPLC, GC, or other analytical techniques.