GSK1120212 ~99.5%

Supplier: 0000018453

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

S2673-5MG S2673-10MG S2673-50MG
101759-296EA 181.54 USD
101759-296 101759-298 101759-300
GSK1120212 ~99.5%
GSK1120212

Trametinib is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays, no inhibition of the kinase activities of c-Raf, B-Raf, ERK1/2.

Formula: C₂₆H₂₃FIN₅O₄
MDL Number: MFCD17215075
CAS Number: 871700-17-3

Order Now

Specification Test Results

DMSO 22 mg/mL warmed (35.74 mM)
Water Insoluble
Ethanol Insoluble
4% DMSO+corn oil 3 mg/mL
Purity (HPLC) 99.65%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR