Net Rings, 20 µm, Cytiva
Supplier: Cytiva
28966379
101725-278EA 147.62 USD
101725-278
Net Rings, 20 µm, Cytiva
Seals Sealing Rings
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions: chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol.
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.