O-Ring, AC 10, Cytiva
Supplier: Cytiva
19504301
101724-990EA 109.62 USD
101724-990
O-Ring, AC 10, Cytiva
Seals Sealing Rings
Resistant to aqueous solutions and solvents used in liquid chromatography.
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.