Nets for C 10 and AC 10, Cytiva
Supplier: Cytiva
19500801
101724-986EA 114 USD
101724-986
Nets for C 10 and AC 10, Cytiva
Seals Sealing Rings
Resistant to aqueous solutions and solvents used in liquid chromatography.
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.