Support Screens for C Columns, Cytiva
Supplier: Cytiva
19176701
101724-974EA 67.23 USD
101724-974
Support Screens for C Columns, Cytiva
Filters Filter Discs
Resistant to aqueous solutions and solvents used in liquid chromatography.
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, 10% HCl, 5% acetic acid, or strong mineral acid.