O-Ring Adapter, Cytiva
Supplier: Cytiva
19016301
101724-952EA 90.62 USD
101724-952
O-Ring Adapter, Cytiva
Chromatography Supplies Chromatography Spare Parts
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions: chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol.
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.