Filter Tool, Cytiva
Supplier: Cytiva
18115320
101724-862EA 249.92 USD
101724-862
Filter Tool, Cytiva
Puller and Inserter Tools
This filter tool is suitable for use with Tricorn columns.
Delivery information: Includes hook and a plunger for changing filters.