TRI CAT Research ELISA
Supplier: ALPCO DIAGNOSTICS MS
17TCTHUE03RES
101721-494EA 3020 USD
101721-494
TRI CAT Research ELISA
Assays ELISAs
The tri-CAT ELISA (Research) is a three-part enzyme immunoassay intended for the quantitative determination of epinephrine, norepinephrine and dopamine in biological fluids and tissue homogenate using small sample volumes.
  • Each catecholamine is measured independently on separate plates
Dopamine is a precursor to epinephrine and norepinephrine, but all three catecholamines act as hormones and/or neurotransmitters. Dopamine acts as a neurotransmitter in the brain, and low levels are often associated with Parkinson's Disease. Dopamine levels may also be increased with tumors of the chromaffine system. Epinephrine and norepinephrine are involved with the response to chronic and acute stress, often termed the 'fight or flight response'.
Caution: For research use only. Not for use in diagnostic procedures.