Norepinephrine Research (Ultrasensitive) ELISA
Supplier: ALPCO DIAGNOSTICS MS
17NORHUE01RES
101721-264EA 1484.62 USD
101721-264
Norepinephrine Research (Ultrasensitive) ELISA
Assays ELISAs
The norepinephrine ELISA (research) is an enzyme immunoassay for the quantitative determination of norepinephrine in biological fluids and tissue homogenate using sample volumes between 1 to 100 ul, depending on sample type and expected concentrations,.
Norepinephrine, epinephrine, and dopamine are members of the catecholamine family and act as hormones and neurotransmitters throughout the body. Along with epinephrine, norepinephrine is involved with the response to chronic and acute stress, often termed the 'fight or flight response' The release of norepinephrine contributes to an increase in heart rate, the release of stored fuels such as fatty acids and glucose for use as energy, and vasoconstriction in the peripheral arteries.
Caution: For research use only. Not for use in diagnostic procedures.