Dopamine ELISA
Supplier: ALPCO DIAGNOSTICS MS
17-DOPHU-E01.1
101721-014EA 1213.85 USD
101721-014
Dopamine ELISA
Assays ELISAs
The dopamine ELISA is an enzyme Immunoassay (ELISA) for the quantitative determination of dopamine in plasma and urine.
Dopamine is extracted by using a cis-diol-specific affinity gel, acylated and then derivatized enzymatically. The competitive ELISA kit uses the microtiter plate format. The antigen is bound to the solid phase of the microtiter plate. The derivatized standards, controls and samples and the solid phase bound analytes compete for a fixed number of antiserum binding sites. After the system is in equilibrium, free antigen and free antigen-antiserum complexes are removed by washing. The antibody bound to the solid phase is detected by an anti-rabbit IgG-peroxidase conjugate using TMB as a substrate. The reaction is monitored at 450 nm.
Caution: For research use only. Not for use in diagnostic procedures.