Alpha 1-Antichymotrypsin ELISA
Supplier: ALPCO DIAGNOSTICS MS
41-A1CHU-E01
101720-684EA 743.08 USD
101720-684
Alpha 1-Antichymotrypsin ELISA
Assays ELISAs
The alpha 1-antichymotrypsin ELISA kit is a highly sensitive two-site enzyme linked immunoassay for measuring alpha 1-antichymotrypsin in biological fluids of humans.
In this assay the alpha 1-antichymotrypsin present in samples reacts with the anti-alpha 1-antichymotrypsin antibodies which have been adsorbed to the surface of polystyrene microtiter wells. After the removal of unbound proteins by washing, anti-CYT antibodies conjugated with horseradish peroxidase (HRP), are added. These enzyme-labeled antibodies form complexes with the previously bound CYT. Following another washing step, the enzyme bound to the immunosorbent is assayed by the addition of a chromogenic substrate, 3,3’,5,5’-tetramethylbenzidine (TMB). The quantity of bound enzyme varies directly with the concentration of CYT in the sample tested; thus, the absorbance, at 450 nm, is a measure of the concentration of CYT in the test sample. The quantity of CYT in the test sample can be interpolated from the standard curve constructed from the standards and corrected for sample dilution.
Caution: For research use only. Not for use in diagnostic procedures.