Histamine ELISA Plasma/Urine
Supplier: ALPCO DIAGNOSTICS MS
17-HISHU-E01
101720-562EA 783.08 USD
101720-562
Histamine ELISA Plasma/Urine
Assays ELISAs
The histamine ELISA is intended for the quantitative determination of histamine in human plasma and urine.
  • Contains two ready-to-use controls
  • Wide standard ranges, convenient measurement of pathological samples without predilution
Histamine belongs to the biogenic amines and is synthesized by decarboxylation from the amino acid histidine. It is synthesized by mast cells, basophils, platelets, histaminergic neurons, and enterochromaffin cells, where it is stored intracellularly in vesicles and released on stimulation. Histamine acts by binding to its four receptors (H1R, H2R, H3R, and H4R) on target cells in various tissues. It causes smooth muscle cell contraction, vasodilation, increased vascular permeability and mucus secretion, tachycardia, alterations of blood pressure, and arrhythmias. Research demonstrates that in humans, histamine is one of the most important mediators and takes part in the initial phase of an anaphylactic reaction ('immediate type' allergy).
Caution: For research use only. Not for use in diagnostic procedures.