Mouse High Range Insulin ELISA
Supplier: ALPCO DIAGNOSTICS MS
80-INSMSH-E01
101720-328EA 496.92 USD
101720-328
Mouse High Range Insulin ELISA
Assays ELISAs
The mouse high range insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes from as little as 5 µl of serum or plasma sample.
  • Low sample volume of 5 µl
  • Broad dynamic range of 3 to 150 ng/ml
  • Superior sensitivity of 1.99 ng/ml
Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the mouse high range insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time. 
Caution: For research use only. Not for use in diagnostic procedures.