Mouse Insulin ELISA
Supplier: ALPCO DIAGNOSTICS MS
80-INSMS-E01
101720-326EA 523.08 USD
101720-326
Mouse Insulin ELISA
Assays ELISAs
The mouse insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes with just 10 µl of serum or plasma sample.
  • Low sample volume of 10 µl
  • Dynamic range of 0.188 to 6.9 ng/ml
  • Superior sensitivity of 0.06 ng/ml
Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the mouse insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time.
Caution: For research use only. Not for use in diagnostic procedures.