Estrone ELISA
Supplier: ALPCO DIAGNOSTICS MS
11-ESRHU-E01
101720-206EA 312.31 USD
101720-206
Estrone ELISA
Assays ELISAs
The Estrone ELISA is a competitive immunoassay for the quantitative determination of estrone in human serum. For in vitro diagnostic use within the United States of America.
  • FDA registered; for in vitro diagnostic use
Estrone is a steroid like estriol and estradiol, belonging to the class of estrogens. The estrogens are involved in the development of female sex organs and secondary sex characteristics. Before the ovum is fertilized, the main action of the estrogens is on the growth and function of the reproductive tract to prepare it for the fertilized ovum. During the follicular phase of the menstrual cycle the estrone level shows a slight increase. The production of estrone then increases markedly to peak at around day 13. The peak is of short duration and by day 16 of the cycle levels will be low. A second peak occurs at around day 21 of the cycle and if fertilization does not occur, then the production of estrone decreases.
Certifications: FDA registered. For in vitro diagnostic use.