NEBNext® Magnesium RNA Fragmentation Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E6150S
101710-296EA 55 USD
101710-296
NEBNext® Magnesium RNA Fragmentation Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext® Magnesium RNA Fragmentation Module has been optimized to fragment RNA into small pieces using divalent cations under elevated temperature
The NEBNext Magnesium RNA Fragmentation Module contains buffers that are suited for RNA fragmentation. Each of these components must pass rigorous quality control standards.