EPIMARK® Methylated DNA Enrichment Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E2600S
101710-274EA 599 USD
101710-274
EPIMARK® Methylated DNA Enrichment Kit, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The EpiMark Methylated DNA Enrichment Kit enables the enrichment of double-stranded CpG methylated DNA based on CpG methylation density.
It utilizes the methyl-CpG binding domain of human MBD2a protein as a capture agent. The protein is fused to the Fc tail of human IgG1 (MBD2a-Fc), which is coupled to Protein A Magnetic Beads (MBD2a-Fc/Protein A Bead). This stable complex will selectively bind double-stranded methylated CpG containing DNA. The high binding affinity of the MBD2a-Fc coupled beads and optimized reagents increases sensitivity and accuracy. This kit contains all the individual components necessary to achieve enrichment in less than two hours using a four step process.