NEBNEXT® dA-Tailing Reaction Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
B6059S
101710-272EA 51 USD
101710-272
NEBNEXT® dA-Tailing Reaction Buffer, New England Biolabs
Buffers Cloning Buffers
New England Biolabs supplies a unique 10X reaction buffer for use with NEBNext® Second Strand Synthesis Enzyme Mix when alternative concentrations of deoxynucleotides are required.