NEBNEXT® QUICK LIGATION™ Reaction Buffer (5X), New England Biolabs
Supplier: New England Biolabs (NEB)
B6058S
101710-268EA 51 USD
101710-268
NEBNEXT® QUICK LIGATION™ Reaction Buffer (5X), New England Biolabs
Buffers Cloning Buffers
New England Biolabs supplies a 5X reaction buffer for use with Quick T4 DNA Ligase
At a 1X concentration this reaction buffer assures optimal activity of the enzyme.