OptiChem® Heating Mantle Controller, Double Circuit, Chemglass
Supplier: Chemglass
CG-15009-01
10170-438EA 794.99 USD
10170-438
OptiChem® Heating Mantle Controller, Double Circuit, Chemglass
Heaters Heating Mantles
OptiChem® line of heating mantle controller.
  • Controllers provide accurate manual control of heating mantles
  • Tapes and all other resistive loads with an adjustable output range from 5 to 100% of the rated voltage
This feature solid-state proportional voltage power control in a compact benchtop design.
Delivery information: Supplied with a 6' separable power cord, a grounded, three wire receptacle and lighted power switch.