1 L Process Filter Reactors, Cylindrical, Unjacketed, Electric, Chemglass
Supplier: Chemglass
CG-1959-U1000
10170-266EA 19206.76 USD
10170-266
1 L Process Filter Reactors, Cylindrical, Unjacketed, Electric, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, and large scale peptide synthesis.
Filter base eliminates material transfers and significantly reducing losses. Complete reactor system on our ChemRxnHub stand. Supplied with the components listed in the link below. 150°C max. temp. limit for filter base.