Richard-Allan™ Parapel™ Paraffin Repellent, Richard-Allan Scientific
Supplier: Epredia
2300
10170-204CS 316.22 USD
10170-204
Richard-Allan™ Parapel™ Paraffin Repellent, Richard-Allan Scientific
Surface Treatments Floor Treatments
Protect against excess buildup of wax on countertops and instrumentation with the Richard-Allan™ Parapel™ Paraffin Repellent.
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations