Microseal® 'C' Optical Seals, Bio-Rad
Supplier: Bio-Rad
MSC1001
101693-716EA 486.15 USD
101693-716
Microseal® 'C' Optical Seals, Bio-Rad
Seals Microplate Seals
Microseal® 'C' Optical Seals are optically clear adhesive films designed for real-time PCR and other optical assays.
The seals provide a tight seal on microplates with raised well rims. Microseal® 'C' Optical Seals are free of DNase, RNase, and human DNA contaminants.
The pressure-sensitive adhesive sticks to the plate, not your gloves.