Anti-IgG Goat Polyclonal Antibody (Cy3®)

Supplier: Genetex
GTX26969
10169-556EA 240.07 USD
10169-556
Anti-IgG Goat Polyclonal Antibody (Cy3®)
Antibodies
Cy3-conjugated Goat anti-Hamster IgG Polyclonal antibody, pre-adsorbed

Type: Secondary
Antigen: IgG antibody, pre-adsorbed
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR