Anti-ARHGEF11 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX16242
10169-498EA 1037.44 USD
10169-498
Anti-ARHGEF11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ARHGEF11

Type: Primary
Antigen: ARHGEF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Chicken, Dog, Monkey, Rat, Chimpanzee, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR