Anti-PHC1 Mouse Monoclonal Antibody [clone: 1F3F3]

Supplier: Genetex
GTX60752
10169-330EA 594.57 USD
10169-330
Anti-PHC1 Mouse Monoclonal Antibody [clone: 1F3F3]
Antibodies
Mouse Monoclonal Antibody [1F3F3] to PHC1

Type: Primary
Antigen: PHC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR